Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner
Tank liner system with top train is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for drums and commonly used industry standard cylindrical tanks. They feature an open top for maximum recovery, with drum dolly available to provide mobility.
775.00€ - 2695.00€
Specifications
Outer Packaging | Cardboard Cartons |
---|---|
Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
Sterilization Method | Irradiated |
Product Line | HyPerforma |
Film Description | CX3-9 |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
17810647
|
Thermo Scientific™
SH30647.10 |
378 L |
815.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17820567
|
Thermo Scientific™
SH30647.11 |
757 L |
2695.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17840567
|
Thermo Scientific™
SH30647.18 |
19 L |
905.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17830567
|
Thermo Scientific™
SH30647.19 |
28 L |
915.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17870567
|
Thermo Scientific™
SH30647.20 |
38 L | N/A | N/A | N/A | |||||
Description
- Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
- Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
- Removes the need for tank cleaning and helps reduce cycle times.
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
Cardboard Cartons | |
Irradiated | |
CX3-9 |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
HyPerforma |