Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Accessories, Empty Wells
These empty 1mL wells are used with Thermo Scientific™ HyperSep 96-well plate applications.
Brand:  Thermo Scientific™ 60300-318
36.35 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10772527
  • 38.30€ / Pack of 100
Only null left
Estimated Shipment: 11-07-2025
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep Empty 1mL Wells are used in HyperSep 96-well plate applications. They are available in fritted or fritless configurations, in packs of 100.
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HyperSep-96 Products and Compatible Manifolds
HyperSep
Empty Wells
1 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Accessories, Empty Wells > 1mL wells