Learn More
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200μL
Get faster sample preparation with minimal sample loss with these SPE micropipette tips and achieve separation in volumes as low as 100nL.
288.00€ - 611.00€
Specifications
For Use With (Application) | Microscale Solid Phase Extraction |
---|---|
For Use With (Equipment) | Micropipettes |
Product Line | HyperSep |
Type | SpinTip |
Volume (Metric) | 10 to 200 ÎĽL |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10404603
|
Thermo Scientific™
60109-412 |
C18; 10-200 uL Vol. | N/A | ||||||
10535435
|
Thermo Scientific™
60109-425 |
Zirconium Dioxide; 10-200 uL Vol. | N/A | ||||||
10140503
|
Thermo Scientific™
60109-422 |
Titanium Dioxide; 10-200 uL Vol. | N/A | ||||||
10138004
|
Thermo Scientific™
60109-421 |
Strong Cation Exchanger; 10-200 uL Vol. | N/A | ||||||
10498684
|
Thermo Scientific™
60109-420 |
Strong Anion Exchanger; 10-200 uL Vol. | N/A | ||||||
10341145
|
Thermo Scientific™
60109-419 |
Weak Anion Exchanger; 10-200 uL Vol. | N/A | ||||||
10426744
|
Thermo Scientific™
60109-418 |
HyperSep SpinTip; 10-200 uL; Trypsin Material | N/A | ||||||
10314673
|
Thermo Scientific™
60109-417 |
HILIC; 10-200 uL Vol. | N/A | ||||||
10716334
|
Thermo Scientific™
60109-416 |
C18 plus Hypercarb; 10-200 uL Vol. | N/A | ||||||
10111273
|
Thermo Scientific™
60109-415 |
Hypercarb; 10-200 uL Vol. | N/A | ||||||
Description
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Features:Ideal for small sample volumes; features a 1 to 2ÎĽm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30ÎĽm).
- Filter-free design to eliminate dead volume
- No contamination from the supporting matrix
- Available in many popular chemistries
- Mass spectrometry
- Desalting
- Protein purification
- MALDI
- Electrophoresis
- HPCE
- HPLC
- CEC
Specifications
Microscale Solid Phase Extraction | |
HyperSep | |
10 to 200 ÎĽL |
Micropipettes | |
SpinTip |