Learn More
STEMCELL Technologies HypoThermosol™ FRS
Animal component-free, defined hypothermic (2 - 8°C) preservation medium for a range of cell and tissue types

414.45€ - 2150.55€
Specifications
Cell Type | Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells |
---|---|
For Use With (Application) | Cryopreservation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109931
![]() |
STEMCELL Technologies
07945 |
500 mL Bag |
2150.55€
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17139921
![]() |
STEMCELL Technologies
07934 |
16 x 10 mL |
1610.55€
Pack of 16 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17149921
![]() |
STEMCELL Technologies
07935 |
100 mL |
414.45€
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17159921
![]() |
STEMCELL Technologies
07936 |
500 mL Bottle |
2103.30€
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- HypoThermosol™ FRS is an optimized hypothermic (2 - 8°C) preservation medium that enables improved and extended preservation of cells, tissues, and organs
- HypoThermosol™ FRS is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process
- HypoThermosol™ FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures
- Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers
Specifications
Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells | |
Cryopreservation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.