missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Ibrutinib Product Code.: 30117158

Thermo Scientific Chemicals Ibrutinib

Product Code. 30117158
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 30117158

Brand: Thermo Scientific Chemicals 466800010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936563-96-1
Molecular Formula C25H24N6O2

Specifications

Quantity 1 g
Formula Weight 440.51
InChI Key XYFPWWZEPKGCCK-UHFFFAOYNA-N
SMILES NC1=C2C(=NC=N1)N(N=C2C1=CC=C(OC2=CC=CC=C2)C=C1)C1CCCN(C1)C(=O)C=C
IUPAC Name 1-{3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]piperidin-1-yl}prop-2-en-1-one
Molecular Weight (g/mol) 440.51
Chemical Name or Material Ibrutinib

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461860 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number466800010,466802500
Lot NumberA0450110 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog Number466800010,466802500
Lot NumberA0443999 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number466800010,466802500
Lot NumberA0445982 Certificate TypeCertificate of Analysis Date06/02/2023 Catalog Number466800010,466802500
Lot NumberA0442494 Certificate TypeCertificate of Analysis Date31/05/2022 Catalog Number466800010,466802500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Ibrutinib >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.