missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS (CD278) Mouse anti-Human , Alexa Fluor 488, Clone: DX29, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 567003
This item is not returnable.
View return policy
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human samples. It is validated for Flow CytometrySpecifications
ICOS (CD278) | |
Monoclonal | |
Alexa Fluor 488 | |
ICOS; DX-29; H4; Inducible T-cell costimulator; AILIM; CVID1 | |
Activated human T cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
DX29 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |