Learn More
ICP-MS Memory Check Standard solution B for 6020 CLP-M, Specpure™, Thermo Scientific Chemicals
213.06€ - 689.43€
Specifications
Chemical Name or Material | ICP-MS Memory Check Standard solution B for 6020 CLP-M |
---|---|
Density | 1.0 g/mL |
MDL Number | MFCD00803323 |
UN Number | UN3264 |
Physical Form | Liquid |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11338468
|
Thermo Scientific Alfa Aesar
040488.AE |
100 mL |
213.06€
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400281
|
Thermo Scientific Alfa Aesar
040488.AP |
500 mL |
689.43€
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
ICP-MS Memory Check Standard solution B for 6020 CLP-M | |
MFCD00803323 | |
Liquid | |
Specpure™ |
1.0 g/mL | |
UN3264 | |
Matrix: 2% HNO3/tr. F-/tr. tartaric acid |
Sicherheit und Handhabung
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
H290-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures