Learn More
WTW™ IDS Conductivity Cell TetraCon™ 925/LV with Fork Design
The WTW™ IDS Conductivity Cell 925/LV with Fork Design has proven TetraCon™ technology and is ideal for small samples such as pastes.
Brand: WTW™ 301719
800.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- IDS conductivity measurement
- Proven four-electrode technology
- Automatic transfer of cell constant and temperature compensation
- Particularly suited for small samples (ca. 2.5 mL) and paste-like samples
- Cell constant 0.469 cm-1
- Fork design
Specifications
IDS conductivity cell with four graphite electrodes, epoxy shaft | |
Small Sample Measurement (2.0.5 mL), Especially Pastes | |
Epoxy Body |
Conductivity Cell | |
Plug head for AS/IDS-x cables or wireless modules IDS WLM-S |
Product Content Correction
Product Title
WTW™ IDS Conductivity Cell TetraCon™ 925/LV with Fork Design > For small sample measurement