Learn More
Novus Biologicals™ IFN-alpha/beta R1 Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-04169
Product Code. 18058606
  • 700.00€ / 0.10mg
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The IFN-alpha/beta R1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_62. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens IFNAR1, mRNA.
Native