missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma Antibody (K3.53), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2852MFV610
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow CytometrySpecifications
IFN-gamma | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
3458 | |
Store at 4C in the dark. |
Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
K3.53 | |
Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
E. coli-derived recombinant human IFN-gamma | |
RUO | |
Primary | |
Human | |
IgG2a |