Learn More
Ig, λ1, λ2 and λ3 Light Chain, Clone: R26-46, Secondary Antibody, BD
Brand: BD Biosciences 553432
266.44 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The R26-46 antibody reacts specifically with mouse Igs bearing λ1, λ2, or λ3 light chains. It does not react with κ light chain or heavy chain. Detection of surface immunoglobulin on Ig λ chain-secreting hybridoma cells has bee
Host Species: Rat
Clone: R26-46
Isotype: IgG2a, Îş
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
ELISA

Specifications
Ig, λ1, λ2 & λ3 Light Chain | |
R26-46 | |
Rat | |
0.5 mg | |
Immunology | |
Mouse |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG2a Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.