Learn More
IgD Mouse anti-Human , BUV615, Clone: IA6-2 (also known as δ-IA6-2), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 613008
309.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow Cytometry
Specifications
IgD | |
Monoclonal | |
Brilliant Ultraviolet 615 | |
IGHD; Ig delta chain C region; Immunoglobulin heavy constant delta | |
Human IgD | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
IA6-2 (also known as δ-IA6-2) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.