missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Fc, Canine, Biotin, Polyclonal, Isotype Control, Novus Biologicals™
Canine Antibody
Brand: Novus Biologicals NBP1-97237-0.5MG
This item is not returnable.
View return policy
Description
IgG Fc specifically detects IgG Fc in Canine samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgG Fc | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay | |
Canine | |
0.5 mg | |
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Canine IgG, anti-Canine IgG F(c) and anti-Canine Serum. No reaction was observed against anti-Canine IgG F(ab)2 or anti-Papain. | |
Canine | |
Lyophilized |
Immunocytochemistry, Immunofluorescence | |
Biotin | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL (BSA) - Immunoglobulin and Protease free | |
Ion exchange chromatography | |
Isotype Control (Primary) | |
Reconstitute with 1.0 mL deionized water. | |
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |