missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Donkey anti-Mouse, Biotin-SP, Secondary Antibody, Proteintech
Donkey anti-Mouse Affinipure Secondary Antibody
50.00€ - 179.00€
Specifications
Antigen | IgG(H+L) |
---|---|
Applications | ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry |
Conjugate | Biotin-SP |
Form | Liquid |
Host Species | Donkey |
Specifications
IgG(H+L) | |
Biotin-SP | |
Donkey | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry | |
Liquid | |
Mouse | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IgG(H+L) Donkey anti-Mouse, Biotin-SP, Secondary Antibody, Proteintech