missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IGH/MALT1 DY Translocation FISH Probe

Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.

1515.00€ - 2495.00€

Specifications

Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Gene immunoglobulin heavy locus
Gene Alias IGH,IGHDY1,MGC72071,MGC88774
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16023575
View Documents
Abnova™
FT0021.100uL
100 μL
1515.00€
100µL
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
16033575
View Documents
Abnova™
FT0021.200uL
200 μL
2495.00€
200µL
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Store at 4°C in the dark
Fluorescent In Situ Hybridization (Cell)
IGH,IGHDY1,MGC72071,MGC88774
3492;10892
Liquid
Tube
immunoglobulin heavy locus
IGH
DNA/RNA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.