Learn More
Proteintech IHCeasy CD8 Ready-To-Use IHC Kit
A ready-to-use IHC kit for the staining of CD8 (CD8A).
Brand:  Proteintech KHC0014-50T
Product Code. 17426723
  • 389.00€ / 1 set
Explore more special offers
Description
Description
  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
Human
Store at 2°C to 8°C
50 Reactions
FFPE Tissue
50 Tests
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Proteintech IHCeasy CD8 Ready-To-Use IHC Kit > 50 Tests