Learn More
Invitrogen™ Human IL-13 ProQuantum Immunoassay Kit
GREENER_CHOICE
ProQuantum Immunoassay Kit
Brand:  Invitrogen™ A35609
 View more versions of this product
Product Code. 15980483
  • 748.00€ / 96 reactions
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
IL-13 Human ProQuantum Immunoassay Kit for ELISA
  • ProQuantum Immunoassay Kit
  • For ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C.
96 Tests