Learn More
Invitrogen™ IL-17 E Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand:  Invitrogen™ 900-M234
Product Code. 17889185
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q9H293
64806
2 x 96 Tests
Refer to product documentation for component specific storage temperature.
ELISA
IL25