missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-3 Human ProQuantum Immunoassay Kit
GREENER_CHOICE

Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument

748.00€ - 3350.00€

Specifications

Accession Number P08700
Assay Range 0.0128 to 5000 pg/mL
Assay Sensitivity <0.01 pg/mL
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15950473
GREENER_CHOICE
View Documents
Invitrogen™
A35586
96 Tests
748.00€
96 reactions
In Stock
to see stock.
Add to basket
 
17103109
GREENER_CHOICE
View Documents
Invitrogen™
A355865
5 x 96 Tests
3350.00€
Each
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
 
Description

Description

  • High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
  • Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
  • Fast, easy workflow: No wash steps, 2 hours from sample to answer
  • Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
  • No proprietary instrument to purchase: Runs on any real-time PCR instrument
  • ProQuantum software available: Robust data analysis and statistical group wise comparison
Specifications

Specifications

P08700
<0.01 pg/mL
Proximity-based Immunoassay
Human
qPCR Instruments
IL-3; MCGF; MULTI-CSF
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard
RUO
Frozen at -20°C
30 min.
0.0128 to 5000 pg/mL
Taqman™ Real-time PCR
ProQuantum
Protein quantitation
3562
1 hr.
FAM™
Serum,2-5μL; Supernatant,2-5μL; Plasma,2-5μL
Human
2 hr.
Videos
SDS
Documents

Documents

Product Certifications