missing translation for 'onlineSavingsMsg'
Learn More

IL-33R (ST2) Rat anti-Mouse, PE, Clone: U29-93, BD

Rat Monoclonal Antibody

144.00€ - 313.00€

Specifications

Antigen IL-33R (ST2)
Clone U29-93
Concentration 0.2mg/mL
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15824939
View Documents
BD Biosciences
566312
25 μg
144.00€
Each
Please to purchase this item. Need a web account? Register with us today!
15814939
View Documents
BD Biosciences
566311
100 μg
313.00€
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The U29-93 monoclonal antibody specifically binds to the mouse Interleukin-33 Receptor (IL-33 Receptor, or IL-33R) which is also known as ST2. The IL-33R exists in either a type I transmembrane or soluble glycoprotein form. These IL-33R forms are encoded by the Il1rl1 (Interleukin-1 receptor-like 1) gene which belongs to the IL-1 Receptor family within the Ig superfamily. The IL-33R is expressed by subsets of T cells, including Th2-like cells and some regulatory T cells, as well as some innate lymphocytes, eosinophils, basophils, and mast cells. The IL-33R (also known as the IL-33R alpha subunit, or binds IL-33 and complexes with the IL-1R Accessory Protein (IL1RAP) to form a functional signaling receptor complex that can induce the production of T helper type 2 (Th2) cytokines. The soluble IL-33R may function as a decoy receptor which can block the binding of IL-33 to the transmembrane IL-33R. The IL-33R plays roles in inflammation, immunity, and allergy.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
Specifications

Specifications

IL-33R (ST2)
0.2mg/mL
PE
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse IL-33R Recombinant Protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
U29-93
Monoclonal
Rat
Mouse
Il1rl1, ST2, ST2L, IL-33R alpha, IL-33Rα, IL33RA, Ly84, DER4, Fit-1
IgG2a κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IL-33R (ST2) Rat anti-Mouse, PE, Clone: U29-93, BD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.