missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-4 Human ProQuantum Immunoassay Kit
GREENER_CHOICE

Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument

Brand:  Invitrogen™ A355875

 View more versions of this product

Product Code. 17113109

  • 3350.00€ / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
  • Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
  • Fast, easy workflow: No wash steps, 2 hours from sample to answer
  • Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
  • No proprietary instrument to purchase: Runs on any real-time PCR instrument
  • ProQuantum software available: Robust data analysis and statistical group wise comparison
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P05112
<0.01 pg/mL
Proximity-based Immunoassay
Human
qPCR Instruments
BCGF-1; BCGF1; BSF-1; BSF1; IL-4
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard
5 x 96 Tests
Serum,2-5μL; Supernatant,2-5μL; Plasma,2-5μL
Human
2 hr.
0.0128 to 5000 pg/mL
Taqman™ Real-time PCR
ProQuantum
Protein quantitation
3565
1 hr.
FAM™
RUO
Frozen at -20°C
30 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers