Learn More
IL-4 Mouse, anti-Human, R718, Clone: MP4-25D2, BD Horizon™
Mouse Monoclonal Antibody
372.00€ - 555.00€
Specifications
Antigen | IL-4 |
---|---|
Clone | MP4-25D2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17207774
|
BD Biosciences
566981 |
25 Tests |
372.00€
25 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
17297764
|
BD Biosciences
566980 |
100 Tests |
555.00€
100 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
The MP4-25D2 monoclonal antibody specifically binds to Interleukin-4 (IL-4). IL-4 is also known as Lymphocyte stimulatory factor 1, B cell stimulatory factor 1 (BSF-1), or B cell growth factor 1 (BCGF-1). IL-4 is produced by activated T cells, basophils, and mast cells. IL-4 is a multifunctional cytokine and growth factor that affects a variety of different target cell types. IL-4 can costimulate T cell proliferation and survival, as well as help drive Th2-like cell differentiation and effector functions. It costimulates B cell proliferation and survival, and can help B cells differentiate into IgG4- or IgE-producing cells. IL-4 can also diminish the proinflammatory functions of monocytes and macrophages. IL-4 exerts its biological effects by binding with high affinity to cell surface CD124 (IL-4Rα chain). CD124 forms signaling IL-4 receptor complexes with either CD132/common γ chain or CD213a1/IL-13Rα1 to form either Type I or Type II IL-4 Receptor complexes, respectively. The immunogen used to generate the MP4-25D2 hybridoma was purified recombinant human IL-4. This is a neutralizing antibody. The MP4-25D2 antibody has been reported to crossreact with IL-4 from rhesus monkeys. The use of the MP4-25D2 antibody for epitope mapping of human IL-4 has been described.
Specifications
IL-4 | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
Refrigerate Upon Arrival |
MP4-25D2 | |
Monoclonal | |
Liquid | |
RUO | |
Primary |