missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Solution dNTPs: DNA Polymerization Mix

Free from DNase, RNase, and nicking enzyme activity

141.11€ - 379.00€

Specifications

Format Liquid
Concentration 20 mM
Purity >99%
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
View More Specs

Products 2
Product Code Brand Concentration Product Type Quantity Price Quantity & Availability  
Product Code Brand Concentration Product Type Quantity Price Quantity & Availability  
10196414
View Documents
Cytiva
28-4065-58
20 mM DNA Polymerase Mix dNTP (20mM each A,C,G,T) 4 x 10 μm
379.00€
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
10793007
View Documents
Cytiva
28-4065-57
20 mM DNA Polymerization Mix dNTP (20mM each A,C,G,T) 10 μm
141.11€
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

Specifications

Specifications

Liquid
>99%
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
20 mM
Colorless
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • dNTP Mix - A C G T, 4 x 10

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-4,8 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 4,8% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Solution dNTPs: DNA Polymerization Mix

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.