Learn More
Thermo Scientific™ Imject™ Maleimide-Activated Blue Carrier™ Protein
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
Brand:  Thermo Scientific™ 77661
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11821184
  • 134.00€ / 2mg
Description
Thermo Scientific™ Maleimide-Activated Blue Carrier Protein is sulfhydryl-reactive immunogenic hemocyanin (CCH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Blue Carrier Protein is a large homocyanin that is highly immunogenic like keyhole limpet hemocyanin (KLH) yet is more soluble. The carrier is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each Blue Carrier Protein molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest.
Highlights:
  • Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of carrier, ensuring consistently high levels of conjugatio
  • Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
  • Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
  • High-quality carrier protein – Blue Carrier Protein is ultrapure Concholepas concholepas hemocyanin (CCH) prepared from molluscan hemolymph
  • Highly immunogenic and soluble – CCH has a high molecular mass that elicits a stronger immune response than BSA or ovalbumin
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Upon receipt store at -20°C. Product is shipped on ice.
2 mg
Blue Carrier™, Imject™
Imject Maleimide-Activated Blue Carrier Protein
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberWE326691 Certificate TypeCertificate of Analysis Date10/06/2021 Catalog Number77661
Lot NumberVC295519 Certificate TypeCertificate of Analysis Date19/03/2020 Catalog Number77661
Lot NumberSD243750 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog Number77661
Lot NumberQE217414 Certificate TypeCertificate of Analysis Date15/04/2017 Catalog Number77661
Lot NumberOL197215 Certificate TypeCertificate of Analysis Date14/04/2017 Catalog Number77661
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Imject™ Maleimide-Activated Blue Carrier™ Protein > Activated Carrier Protein; 2mg