Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™ 1004170
 View more versions of this product
Product Code. 11726188
  • 45.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Description
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Specifications
Mesh Basket
Ultrasonic Cleaner, Elma E, S, P 10 Series Benchtop 0.8 L
Stainless Steel
Videos
  • Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
    Product Title
    Fisherbrand™ Immersion Basket with Handle > For E, S, P 10 Series Benchtop Ultrasonic Cleaners