Learn More
STEMCELL Technologies ImmunoCult™-ACF Dendritic Cell Medium
Medium for the culture and differentiation of human monocytes into dendritic cells and their maturation
Brand:  STEMCELL Technologies 10986
Encompass
 View more versions of this product
Product Code. 16655635
  • 723.00€ / 500mL
Estimated Shipment: 20-11-2024
Description
  • ImmunoCult™-ACF Dendritic Cell Medium and Supplements, sold individually or as a kit, have been developed for the in vitro culture and differentiation of human monocytes into dendritic cells (DCs)
  • ImmunoCult™-ACF Dendritic Cell Medium is a serum-free and animal component-free medium optimized for the in vitro culture and differentiation of human monocytes into DCs
  • ImmunoCult™-ACF Dendritic Cell Differentiation Supplement contains a combination of animal component-free recombinant human cytokines formulated to support the differentiation of immature DCs from human monocytes
  • The ImmunoCult™ Dendritic Cell Maturation Supplement has been formulated to support maturation of immature DCs to mature DCs
Specifications
Specifications
Dendritic Cells, Monocytes
500 mL