missing translation for 'onlineSavingsMsg'
Learn More
Indole-5-carboxaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 11437196

Indole-5-carboxaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11437196
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11437196

Brand: Thermo Scientific Alfa Aesar B25517.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1196-69-6 | C9H7NO | 145.161 g/mol

Indole-5-carboxaldehyde is used as a reactant in preparation of curcumin derivatives as anti-proliferative & anti-inflammatory agents; and in synthesis of para-para stilbenophanes by McMurry coupling. It is also used as a reactant in stereoselective synthesis of dibenzylideneacetone derivatives as β-amyloid imaging probes and in structure-based drug design of aurora kinase A inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-5-carboxaldehyde is used as a reactant in preparation of curcumin derivatives as anti-proliferative & anti-inflammatory agents; and in synthesis of para-para stilbenophanes by McMurry coupling. It is also used as a reactant in stereoselective synthesis of dibenzylideneacetone derivatives as β-amyloid imaging probes and in structure-based drug design of aurora kinase A inhibitors.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1196-69-6
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD02093664
InChI Key ADZUEEUKBYCSEY-UHFFFAOYSA-N
Synonym indole-5-carboxaldehyde, 5-formylindole, indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 5-indolealdehyde, indole-5-aldehyde, 5-formyl-1h-indole, 5-indolecarboxaldehyde, indole-5-carboxaldehyde 5-formylindole, 5-formyl-indole
PubChem CID 589040
IUPAC Name 1H-indole-5-carbaldehyde
SMILES C1=CC2=C(C=CN2)C=C1C=O

Specifications

Melting Point 96°C to 100°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material Indole-5-carboxaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10237403 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB25517.06,B25517.03
Lot Number10163161 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25517.06,B25517.03
Lot Number10160862 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25517.06,B25517.03
Lot Number10195840 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25517.06,B25517.03
Lot Number10161910 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25517.06,B25517.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indole-5-carboxaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.