Learn More
B Braun™ Injekt Solo Cone Syringes
Enjoy the smooth, steady operation of the B Braun™ Solo Cone Leur syringe. Its green plunger enables easy recognition of filling volumes and its double-seal stopper keeps contents intact.
Brand:  B Braun™ 9202625
 View more versions of this product
Product Code. 10542344
  • 123.00€ / Pack of 1000
Description
Enjoy the smooth, steady operation of the B Braun™ Solo Cone Leur syringe. Its green plunger enables easy recognition of filling volumes and its double-seal stopper keeps contents intact.
  • Injekt™
  • Transparent barrel with graduations
  • Green plunger
  • Barrel and plunger rod: Polypropylene
  • No DEHP or natural rubber
Specifications
Specifications
EN ISO 7886-1
Single-use
10 mL
Syringe
1/Ea.
Non-sterile
Certificates
Promotions
Promotions
Product Content Correction
Product Title
B Braun™ Injekt Solo Cone Syringes > Capacity: 20mL, Non-Sterile