Learn More
Mettler Toledo™ InLab™ 605 Dissolved Oxygen Sensor
Brand:  Mettler Toledo™ 51340292
1140.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11957761
  • 1170.00€ / Each
Estimated Shipment: 24-06-2025
Specifications
10 m
0°C to 60°C
CE, ISO 9001 certified
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Mettler Toledo™ InLab™ 605 Dissolved Oxygen Sensor > Length Cable: 10m