Learn More
Mettler Toledo™ InLab™ Trace Kit
Brand:  Mettler Toledo™ 30014099
2870.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12946387
  • 2945.00€ / 1 set
Specifications
Trace Kit
0°C to 100°C
1.8 m
Product Title
Mettler Toledo™ InLab™ Trace Kit > Cable Length: 1.8m