Learn More
Abeba™ Insoles 3566
Massage knobs stimulate circulation. ESD dissipative and antistatic.
13.63€ - 17.45€
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15610208
|
Abeba™
3566/40 |
40 |
17.45€
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
15620208
|
Abeba™
3566/41 |
41 |
17.45€
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
15630208
|
Abeba™
3566/42 |
42 |
17.45€
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
15640208
|
Abeba™
3566/43 |
43 | N/A | |||||||
15650208
|
Abeba™
3566/44 |
44 |
17.45€
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
15660208
|
Abeba™
3566/45 |
45 |
17.45€
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
15670208
|
Abeba™
3566/46 |
46 |
17.45€
1 pair |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
Description
- Microfiber
- ESD dissipative according to DIN EN 61340
- Antistatic
- Stimulating circulation by massage knobs, comfortable orthopaedic footbed
- Reflexor massage active-footbed
Specifications
Microfiber | |
3566 |
Product Title
Abeba™ Insoles 3566