Learn More
Micronova™ Inspection Mirror with Plastic Bottom Plate
An electropolished stainless steel mirror with a swivel joint and handle connection for inspecting hard to reach areas.
Brand:  Micronova™ QDIST-66P
334.40 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Product Code. 15712819
  • 352.00€ / Each
Estimated Shipment: 15-05-2025
Description
  • A highly polished stainless steel mirror for inspecting hard to reach areas
  • Features a swivel connection allowing it to be attached to fixed and extension handles
  • This allows the operator to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned
  • The optional flashlight attachment is ideal for use in tanks and other low light applications
  • QD versions connect with SSU series stainless steel handles
  • Certifications
    Certificate of conformance
    Certificates
    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number047737-000 Certificate TypeCertificate of Compliance Date19/02/2025 Catalog NumberQDIST-66P
    Lot Number047306-000 Certificate TypeCertificate of Compliance Date31/12/2024 Catalog NumberQDIST-66P
    Lot Number047214-000 Certificate TypeCertificate of Compliance Date31/12/2024 Catalog NumberQDIST-66P
    Lot Number046993-000 Certificate TypeCertificate of Compliance Date16/12/2024 Catalog NumberQDIST-66P
    Lot Number045241-000 Certificate TypeCertificate of Compliance Date11/11/2024 Catalog NumberQDIST-66P
    Product Title
    Micronova™ Inspection Mirror with Plastic Bottom Plate > Material: Stainless Steel, Polypropylene