Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Click to view available options
Range:
0 to 500 mg/L Total
0.00 to 10.00 mg/L Free and Total
0.00 to 5.00 mg/L Total
0.000 to 0.500 mg/L Free
No. of Tests:
100
300
600
For Use With (Application):
Chlorine Ultra High Range
Free Chlorine Ultra Low Range
Free and Total Chlorine High Range
Total Chlorine
Total Chlorine (DPD3)
Total Chlorine Ultra Low Range
Unit Size:
Each
Pack of 100
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
- Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
Specifications
| For Use With (Application) | Free Chlorine Ultra Low Range |
| For Use With (Equipment) | Compatible Hanna benchtop and portable photometers |
| Includes | 100 individual packets of HI95762-0 |
| No. of Tests | 100 |
| Quantity | 1 Set |
| Range | 0.000 to 0.500 mg/L Free |
| Test Method | Standard Method 4500-Cl G |
| Type | Water Testing Kit |