Learn More
Lauda™ Insulation Tube
Manufactured from silicone. Lauda™ Insulation Tube is suitable for hoses with max outer diameter.
Brand:  Lauda™ RKJ 024
Product Code. 10383722
  • 25.65€ / 1m
Estimated Shipment: 18-02-2025
Description
  • For hoses with max outer diameter
Specifications
Specifications
105°C
EPDM Rubber
105°C
16 mm