missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ INT 131
PPARÎł partial agonist
173.00€ - 661.00€
Specifications
Chemical Name or Material | 2,4-Dichloro-N-[3,5-dichloro-4-(3-quinolinyloxy)phenyl]benzenesulfonamide |
---|---|
CAS | 315224-26-1 |
Target | PPAR gamma/NR1C3 |
Molecular Formula | C21H12Cl4N2O3S |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18709696
|
Tocris Bioscience™
7423/10 |
10 mg |
173.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18716706
|
Tocris Bioscience™
7423/50 |
50 mg |
661.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INT 131 is a potent and selective PPARÎł partial agonist (EC50 = 4 nM; Ki = 3.7 nM). It has a 20-fold higher affinity for PPARÎł compared with that of Rosiglitazone (Cat. No. 5325). INT 131 improves glucose tolerance in a rodent model of diabetes without inducing hemodynamic and cardiovascular effects. Antidiabetic agent.Specifications
2,4-Dichloro-N-[3,5-dichloro-4-(3-quinolinyloxy)phenyl]benzenesulfonamide | |
PPAR gamma/NR1C3 | |
0.98 |
315224-26-1 | |
C21H12Cl4N2O3S |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ INT 131