missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Mouse anti-Human, Clone: MEM48, Novus Biologicals™
Mouse Monoclonal Antibody
530.00€ - 1065.00€
Specifications
Antigen | Integrin beta 2/CD18 |
---|---|
Clone | MEM48 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392552
|
Bio-Techne
NBP3-14608-200UL |
200 μg |
530.00€
200µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18353233
|
Bio-Techne
NBP3-14608-500UL |
500 μg |
1065.00€
500µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional AssaySpecifications
Integrin beta 2/CD18 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay | |
Unconjugated | |
Mouse | |
Apoptosis, Cytokine Research, Immunology | |
PBS. Sterile-filtered through 0.22 um. | |
3689 | |
IgG1 κ | |
Affinity purified | |
Antibody |
MEM48 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional | |
Monoclonal | |
Purified | |
RUO | |
Human | |
959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
Leucocytes from an LGL - type leukaemia | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title