Learn More
Diversey™ IntelliCare Dispenser Hybrid
The IntelliCare Hybrid dispensing system can address all hand care needs with a versatile, modern, and durable platform that is compatible with both liquid/gels and foam products.
136.00€
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
17663105
|
Diversey™
D7524179 |
Black |
136.00€
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
17673105
|
Diversey™
D7524180 |
White |
136.00€
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
- Dispenses 1300 mL bottles of hand soap and 1200 mL of alcohol based hand sanitizer
- Can dispense 0.4 mL or 1.00 mL per push
- Hybrid dispenser provides an automatic switch from touchless to manual if batteries died before replacement
- Near empty product alarm and low battery signal
- Locking dispenser contains a translucent window for product visibility without product tampering
Specifications
Hybrid Dispenser (Manual/Automatic) |
Product Title
Diversey™ IntelliCare Dispenser Hybrid