missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Interleukin 2, human
24.30€ - 688.43€
Specifications
Molecular Weight (g/mol) | 15.5 kd |
---|---|
Packaging | Plastic bottle |
Form | Lyophilized |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438699
|
Thermo Scientific Chemicals
|
10μg | N/A | ||||||
15448699
|
Thermo Scientific Chemicals
J67298.I& |
300.50€
50µg |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||||
15458699
|
Thermo Scientific Chemicals
J67298.M^ |
1mg |
688.43€
1mg |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
15.5 kd | |
Plastic bottle | |
Lyophilized |
RUO – Research Use Only