missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Interleukin 3, mouse

136.68€ - 217.00€

Specifications

Molecular Weight (g/mol) 15.6 kd
Packaging Plastic bottle
Form Lyophilized
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428639
View Documents
Thermo Scientific Chemicals
J67104.&E
2μg
136.68€
2µg
Please to purchase this item. Need a web account? Register with us today!
15418639
View Documents
Thermo Scientific Chemicals
J67104.!E
10μg N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

15.6 kd
Plastic bottle
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Interleukin 3, mouse

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.