Learn More
Iodine Solution, 0.05M, Chem-Lab
Product Code. 11903163
Click to view available options
Quantity:
500 mL
1 L
Used in laboratories for accurate volumetric analysis and titrations. Iodine Solution, 0.05M, Chem-Lab is a standardized solution with 12.69g I2 per 1L water. It is also used topically as a disinfectant and antibacterial agent. (Calibrate before use.)
Iodine is a nonmetallic element of the halogen group. It is a nutritionally essential element, especially important in thyroid hormone synthesis. In solution, it has anti-infective properties (when used topically).
Chemical Identifiers
CAS 7553-56-2, 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II
Specifications
pH 5.8–6.2
Quantity 500 mL
IUPAC Name diiodine
Formula Weight 253.81g/mol
Concentration or Composition (by Analyte or Components) 0.05mol/L
Density 1.02g/mL