Learn More
Iodine Solution, Ph. Eur. Grade, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Specifications
Specifications
Quantity | 1 L |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
I00525A1 | Certificate of Analysis | 18/02/2025 | 3002700 |
I00524K1 | Certificate of Analysis | 07/01/2025 | 3002700 |
I00524G1 | Certificate of Analysis | 27/08/2024 | 3002700 |
I00524D1 | Certificate of Analysis | 28/05/2024 | 3002700 |
I00523J1 | Certificate of Analysis | 21/11/2023 | 3002700 |
Product Content Correction
Product Title
Iodine Solution, Ph. Eur. Grade, Reagecon™ >