Learn More
Memmert™ IQ/OQ Document with Test Data for UF55plus
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand:  Memmert™ UF55plusD00127
Product Code. 17589485
  • 2175.00€ / Each
Explore more special offers
Description
Description
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
Specifications
UF55plus Oven
IQ/OQ Documentation
Test Data
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ IQ/OQ Document with Test Data for UF55plus > For Use With: UF55plus