Learn More
Grant Instruments™ IQ/OQ Documentation
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ IQ/OQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
238.50€ - 705.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15271186
|
Grant Instruments™
IQOQ QBH |
All QBH Block Heater Models |
705.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15211196
|
Grant Instruments™
IQOQ R4 |
Grant R4 Refrigerated Baths/Circulators | N/A | ||||||
15221196
|
Grant Instruments™
IQOQ SAP |
JBA and SBB Unstirred Baths |
562.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15251196
|
Grant Instruments™
IQOQ SUB AQUA PLUS |
All SUB Aqua Plus Models | N/A | ||||||
15211186
|
Grant Instruments™
IQOQ BTD |
All Grant BTD Block Heater Models |
248.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15291196
|
Grant Instruments™
IQOQ XUB |
All Grant XUB Digital Ultrasonic Baths |
705.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15261186
|
Grant Instruments™
IQOQ QBD |
All QBD Block Heater Models |
705.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15231186
|
Grant Instruments™
IQOQ JBN JBA |
JBN/JBA Baths |
562.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15241196
|
Grant Instruments™
IQOQ SBB AQUA PLUS |
All SBB Aqua Plus Models |
562.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15281196
|
Grant Instruments™
IQOQ TX150 |
Grant TX150 Stirred Thermostatic Bath/Circulator |
705.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15201206
|
Grant Instruments™
IQOQ XUBA |
All Grant XUBA Analogue Ultrasonic Baths |
562.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15102625
|
Grant Instruments™
IQOQ CS200G |
CS200G Stirred Thermostatic Bath/Circulator | N/A | ||||||
15182615
|
Grant Instruments™
IQOQ CXG |
CXG Stirred Thermostatic Bath/Circulator | N/A | ||||||
15122615
|
Grant Instruments™
IQOQ T100 |
Grant T100 Stirred Thermostatic Bath/Circulator |
562.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15142615
|
Grant Instruments™
IQOQ TC120 |
Grant TC120 Stirred Thermostatic Bath/Circulator |
705.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15162615
|
Grant Instruments™
IQOQ TXF200 |
Grant TXF200 Stirred Thermostatic Bath/Circulator |
705.00€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15410484
|
Grant Instruments™
IQOQ CVP-2 |
Grant CVP-2 PCR Plate Centrifuge |
464.28€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
15440484
|
Grant Instruments™
IQOQ PHMP-100 |
Grant PHMP-100 Thermoshaker for Microplates |
238.50€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
15480474
|
Grant Instruments™
IQOQ TS-DW |
Grant TS-DW Deep Well Plate Thermoshaker |
238.50€
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
Description
- Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
- Operational Qualification (OQ): instrument is functioning in accordance with its specification
- Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
- It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.