missing translation for 'onlineSavingsMsg'
Learn More
Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Honeywell Fluka™ Product Code.: 15603310

Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Honeywell Fluka™

Product Code. 15603310 Shop All Honeywell Fluka Products
1 kg, Poly Bottle
Click to view available options
Quantity:
500 g
1 kg
25 kg
6 x 1 kg
Packaging:
Paper / Poly Bag
Poly Bottle
This item is not returnable. View return policy

Product Code. 15603310

Brand: Honeywell-Fluka 312351KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., 21-23% Fe basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-10-7, 7664-93-9
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00011007
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
Synonym Ferric sulfate hydrate
PubChem CID 167265
IUPAC Name iron(3+);trisulfate;hydrate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 98.0000%, 2.0000%
Quantity 1 kg
Linear Formula Fe2(SO4)3 · xH2O
UN Number UN3260
Formula Weight 399.88
Percent Purity 21 to 23% (Fe basis)
Grade puriss. p.a.
Packaging Poly Bottle
Chemical Name or Material Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis
compliance-icons compliance-icons
Product Identifier
  • Iron(III) sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iron(III) sulfate hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.