Learn More
Micronova™ Irradiated Sharpie Ultra Fine Markers
Irradiated, ultra fine point, capped permanent marker with fade and water resistant, quick drying, non-toxic ink for use in aseptic environments.
Brand: Micronova™ PEN-61IR
Description
Micronova Irradiated Sharpie Ultra Fine Marker is the original pen style, industry standard permanent marker with an ultra fine point. This cleanroom sharpie marks permanently on most surfaces with quick drying, non-toxic, alcohol based ink that is fade and water resistant to further extend multiple project uses.
- Ultra fine point capped sharpie marker
- Marks permanently on most surfaces
- Writes on cold or wet surfaces, plastic bags and disposable lab-ware
- Nontoxic, permanent, quick drying, alcohol-based ink
- Gamma irradiated for aseptic ISO 5 and other clean production areas
- Fade and water resistant
- Available red, blue or black inks
Specifications
HDPE | |
Black | |
HDPE | |
6 x 0.5 x 0.5 in. (15.24 x 1.27 x 1.27 cm) | |
12.24 cm | |
1.27 cm | |
ISO 9001 Certified Facility; Certificate of Irradiation Processing; Certificate of Conformance | |
1.27 cm | |
Cleanroom labels and documents | |
Individually bagged, 4 pens per double bagged pack, 3 packs per case | |
12/Cs. | |
Sterile |
No | |
Black | |
Ink does not smear when used on labels that are autoclaved | |
6 in. | |
0.5 in. | |
Not Autoclavable | |
0.5 in. | |
Reusable | |
Cap | |
Smear-Proof Marker | |
8 x 8 x 8 in. | |
Fiber |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
046235-000 | Certificate of Analysis | 09/12/2024 | PEN-61IR |
044635-000 | Certificate of Quality | 26/08/2024 | PEN-61IR |
044259-000 | Certificate of Quality | 08/07/2024 | PEN-61IR |
042898-000 | Certificate of Quality | 03/06/2024 | PEN-61IR |
042037-000 | Certificate of Quality | 05/02/2024 | PEN-61IR |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02