missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRX6 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | IRX6 |
---|---|
Concentration | 800 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17211376
|
Proteintech
24440-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
17201376
|
Proteintech
24440-1-AP-150UL |
150 μL |
399.00€
150µL |
In Stock
Log in to see stock. |
|||||
Description
The IRX6 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human IRX6. This antibody recognizes human, mouse antigen. The IRX6 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Specifications
IRX6 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Rabbit | |
79190 |
800 μg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse | |
Primary |