Learn More
Brady™ ISO Safety Sign 'Wear Laboratory Coat,' French
Safety Sign to signify that a laboratory coat must be worn.
Brand:  Brady™ 311400
 View more versions of this product
Product Code. 17891755
  • 34.60€ / Each
Estimated Shipment: 29-01-2025
Description
  • Provide visual warning that laboratory coat must be worn in the area
Specifications
Specifications
M/M059/FR797/PP-297X420-1
1 Sign
Surface Mount
16.54 in.
11.69 in.
Rectangular
French
Wear Laboratory Coat
White on Blue
Polypropylene
16.54 x 11.69 in.
Facility Identification, Safety Identification
1.55 mm
-40°C to +120°C
-40°F to +248°F
420 x 297 mm
Safety Sign