Learn More
Brady™ ISO Safety Sign without Text - Warning: Low Temperature/Freezing Conditions
Safety sign for hazard identification.
6.20€ - 9.65€
Specifications
Finish | Gloss |
---|---|
Material | Laminated Polyester |
Adhesive Type | Permanent |
Shape | Triangular |
Color | Black on Yellow |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
15772652
|
Brady™
827797 |
43 x 50 mm |
6.20€
Each |
In Stock
Log in to see stock. |
||||||
15871537
|
Brady™
827799 |
87 x 100 mm |
9.65€
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
||||||
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
Gloss | |
Permanent | |
Black on Yellow |
Laminated Polyester | |
Triangular |
Product Title
Brady™ ISO Safety Sign without Text - Warning: Low Temperature/Freezing Conditions