Learn More
Brady™ ISO Square Polypropylene Safety Sign 'Fire Alarm Flashing Light'
ISO Safety Sign displaying flashing fire alarm and flames.
Brand:  Brady™ 311224
 View more versions of this product
Product Code. 17831745
  • 8.75€ / Each
Only null left
Estimated Shipment: 01-07-2025
Description
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Specifications
Class C
Gloss
1 Sign
Facility Identification, Safety Identification
5.91 in.
Square
Universal
Fire Alarm Flashing Light
Photoluminescent on Red
F/F018/NT/PP-PHOLUMC-150X150-1
Polypropylene
Surface Mount
5.91 in.
-40°C to +66°C
-40°F to +150°F
150 x 150 mm
Safety Sign
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ ISO Square Polypropylene Safety Sign 'Fire Alarm Flashing Light' > ISO Safety Sign Fire Alarm; Color: Photoluminescent on Red; Dimensions (L x W): 150 x 150 mm; Certifications/Compliance: Class C