Learn More
KGW Isotherm™ Isotherm Borosilicate Glass Insulating Lid
39.25€ - 51.20€
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | Flexible Applications In Laboratory |
No. per Pack | 1 |
Product Type | Insulating Lid |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10566232
|
KGW Isotherm™
17211 |
1000 mL |
42.10€
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
10660872
|
KGW Isotherm™
17220 |
2500 mL |
51.20€
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
10208991
|
KGW Isotherm™
17222 |
3000 mL |
51.20€
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
10556232
|
KGW Isotherm™
17214 |
1500 mL |
42.10€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10228941
|
KGW Isotherm™
17218 |
2000 mL |
39.25€
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
Specifications
Borosilicate Glass | |
Flexible Applications In Laboratory | |
1 | |
Insulating Lid |
Product Title
KGW Isotherm™ Isotherm Borosilicate Glass Insulating Lid